Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCTTTTCTGAATTGAGAATCA[C/T]TGCTAACTTCAGCTTTCAAACAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600221 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEK - TEK receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000459.4 | Intron | NP_000450.2 | ||||
NM_001290077.1 | Intron | NP_001277006.1 | ||||
NM_001290078.1 | Intron | NP_001277007.1 | ||||
XM_005251561.1 | Intron | XP_005251618.1 | ||||
XM_005251563.1 | Intron | XP_005251620.1 |