Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCGATGATGACCACGAGGATGC[C/T]TATCACCAGCGCCTCTGTTCCTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602974 | ||||||||||||||||||||
Literature Links: |
AQP7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AQP7 - aquaporin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170.2 | Intron | NP_001161.1 | ||||
NM_001318156.1 | Intron | NP_001305085.1 | ||||
NM_001318157.1 | Intron | NP_001305086.1 | ||||
NM_001318158.1 | Intron | NP_001305087.1 | ||||
XM_005251453.3 | Intron | XP_005251510.1 | ||||
XM_006716765.3 | Intron | XP_006716828.1 | ||||
XM_011517866.2 | Intron | XP_011516168.1 | ||||
XM_011517867.2 | Intron | XP_011516169.1 | ||||
XM_017014700.1 | Intron | XP_016870189.1 | ||||
XM_017014701.1 | Intron | XP_016870190.1 | ||||
XM_017014702.1 | Intron | XP_016870191.1 | ||||
XM_017014703.1 | Intron | XP_016870192.1 | ||||
XM_017014704.1 | Intron | XP_016870193.1 | ||||
XM_017014705.1 | Intron | XP_016870194.1 | ||||
XM_017014706.1 | Intron | XP_016870195.1 |