Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCACACTGTAAACAGCAGTAAG[C/T]CTGGACACAATATATGACACTAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604366 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAI1 - dynein axonemal intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281428.1 | Intron | NP_001268357.1 | ||||
NM_012144.3 | Intron | NP_036276.1 | ||||
XM_006716758.3 | Intron | XP_006716821.1 | ||||
XM_011517846.1 | Intron | XP_011516148.1 | ||||
XM_011517847.2 | Intron | XP_011516149.1 | ||||
XM_011517848.1 | Intron | XP_011516150.1 | ||||
XM_011517849.1 | Intron | XP_011516151.1 | ||||
XM_011517850.2 | Intron | XP_011516152.1 | ||||
XM_017014625.1 | Intron | XP_016870114.1 |
FAM219A - family with sequence similarity 219 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |