Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCTTTCGGTAGAGCTGAGTACA[A/G]CAGGCAGTGCTGGGTGGCAGTAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604833 MIM: 600939 | ||||||||||||||||||||
Literature Links: |
CCL27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCL27 - C-C motif chemokine ligand 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006664.3 | 158 | Silent Mutation | TGC,TGT | C,C 33 | NP_006655.1 |
IL11RA - interleukin 11 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142784.2 | 158 | Intron | NP_001136256.1 |
LOC730098 - uncharacterized LOC730098 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |