Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGATGTTGCGCGAGGAAAATGCG[A/G]GACGCCCAGGTCGGTGCTCGGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600712 MIM: 610404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNRNPK - heterogeneous nuclear ribonucleoprotein K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318186.1 | 143 | Intron | NP_001305115.1 | |||
NM_001318187.1 | 143 | Intron | NP_001305116.1 | |||
NM_001318188.1 | 143 | Intron | NP_001305117.1 | |||
NM_002140.4 | 143 | Intron | NP_002131.2 | |||
NM_031262.3 | 143 | Intron | NP_112552.1 | |||
NM_031263.3 | 143 | Intron | NP_112553.1 | |||
XM_005251960.2 | 143 | Intron | XP_005252017.1 | |||
XM_005251963.3 | 143 | Intron | XP_005252020.1 | |||
XM_005251965.2 | 143 | Intron | XP_005252022.1 | |||
XM_011518616.1 | 143 | Intron | XP_011516918.1 | |||
XM_017014668.1 | 143 | Intron | XP_016870157.1 | |||
XM_017014669.1 | 143 | Intron | XP_016870158.1 |
RMI1 - RecQ mediated genome instability 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024945.2 | 143 | UTR 5 | NP_079221.2 | |||
XM_005252211.2 | 143 | UTR 5 | XP_005252268.1 | |||
XM_005252213.2 | 143 | UTR 5 | XP_005252270.1 | |||
XM_011519034.2 | 143 | UTR 5 | XP_011517336.1 | |||
XM_017015140.1 | 143 | Intron | XP_016870629.1 |