Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTACAGGCATAATGGTTATGAA[A/G]GCTTCTGTAGATGATGACGATTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
NAA35 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAA35 - N(alpha)-acetyltransferase 35, NatC auxiliary subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321881.1 | 363 | Silent Mutation | AAA,AAG | K,K 4 | NP_001308810.1 | |
NM_001321882.1 | 363 | Silent Mutation | AAA,AAG | K,K 4 | NP_001308811.1 | |
NM_024635.3 | 363 | Silent Mutation | AAA,AAG | K,K 4 | NP_078911.3 | |
XM_005252127.3 | 363 | Silent Mutation | AAA,AAG | K,K 4 | XP_005252184.1 | |
XM_011518903.2 | 363 | Intron | XP_011517205.1 | |||
XM_011518904.2 | 363 | Intron | XP_011517206.1 | |||
XM_017015012.1 | 363 | UTR 5 | XP_016870501.1 |