Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTATCAGGTAGAAGATACTTT[C/T]CTCTGTTAATAACTAGCCTTGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609386 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMC5 - structural maintenance of chromosomes 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015110.3 | Intron | NP_055925.2 | ||||
XM_005251837.1 | Intron | XP_005251894.1 | ||||
XM_005251838.1 | Intron | XP_005251895.1 | ||||
XM_005251839.1 | Intron | XP_005251896.1 | ||||
XM_017014507.1 | Intron | XP_016869996.1 | ||||
XM_017014508.1 | Intron | XP_016869997.1 | ||||
XM_017014509.1 | Intron | XP_016869998.1 | ||||
XM_017014510.1 | Intron | XP_016869999.1 |
SMC5-AS1 - SMC5 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |