Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCAGGCTGTCCACCCCCATCCT[C/G]AGCCTTCCACACTCTTCTCCGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601965 MIM: 605263 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
S1PR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
S1PR3 - sphingosine-1-phosphate receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHC3 - SHC adaptor protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016848.5 | 8298 | UTR 3 | NP_058544.3 | |||
XM_011518785.2 | 8298 | UTR 3 | XP_011517087.1 | |||
XM_011518786.2 | 8298 | UTR 3 | XP_011517088.1 | |||
XM_017014809.1 | 8298 | Intron | XP_016870298.1 | |||
XM_017014810.1 | 8298 | Intron | XP_016870299.1 |