Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAGCACCTAATTTTTTAAGTCTT[A/T]GTTCCTGTCACAGGCCCACTTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607693 MIM: 601866 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SECISBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SECISBP2 - SECIS binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282688.1 | 2999 | Intron | NP_001269617.1 | |||
NM_001282689.1 | 2999 | Intron | NP_001269618.1 | |||
NM_001282690.1 | 2999 | Intron | NP_001269619.1 | |||
NM_024077.4 | 2999 | Intron | NP_076982.3 | |||
XM_005252196.2 | 2999 | Intron | XP_005252253.1 | |||
XM_005252202.3 | 2999 | Intron | XP_005252259.1 | |||
XM_006717282.2 | 2999 | Intron | XP_006717345.1 | |||
XM_011519000.2 | 2999 | Intron | XP_011517302.1 | |||
XM_011519001.1 | 2999 | Intron | XP_011517303.1 | |||
XM_011519002.1 | 2999 | Intron | XP_011517304.1 | |||
XM_011519003.1 | 2999 | Intron | XP_011517305.1 | |||
XM_017015122.1 | 2999 | Intron | XP_016870611.1 | |||
XM_017015123.1 | 2999 | Intron | XP_016870612.1 | |||
XM_017015124.1 | 2999 | Intron | XP_016870613.1 | |||
XM_017015125.1 | 2999 | Intron | XP_016870614.1 |
SEMA4D - semaphorin 4D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142287.1 | 2999 | UTR 3 | NP_001135759.1 | |||
NM_006378.3 | 2999 | Intron | NP_006369.3 | |||
XM_005251654.3 | 2999 | Intron | XP_005251711.1 | |||
XM_011518123.2 | 2999 | Intron | XP_011516425.1 | |||
XM_011518124.2 | 2999 | Intron | XP_011516426.1 | |||
XM_011518125.1 | 2999 | Intron | XP_011516427.1 | |||
XM_011518127.2 | 2999 | Intron | XP_011516429.1 | |||
XM_011518128.2 | 2999 | Intron | XP_011516430.1 | |||
XM_011518129.1 | 2999 | Intron | XP_011516431.1 | |||
XM_011518130.2 | 2999 | Intron | XP_011516432.1 | |||
XM_011518131.2 | 2999 | Intron | XP_011516433.1 | |||
XM_011518133.2 | 2999 | Intron | XP_011516435.1 | |||
XM_011518134.2 | 2999 | Intron | XP_011516436.1 | |||
XM_017014193.1 | 2999 | Intron | XP_016869682.1 | |||
XM_017014194.1 | 2999 | Intron | XP_016869683.1 | |||
XM_017014195.1 | 2999 | Intron | XP_016869684.1 | |||
XM_017014196.1 | 2999 | Intron | XP_016869685.1 | |||
XM_017014197.1 | 2999 | Intron | XP_016869686.1 | |||
XM_017014198.1 | 2999 | Intron | XP_016869687.1 | |||
XM_017014199.1 | 2999 | Intron | XP_016869688.1 |