Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATAGAAGAAAGTTGGCCTTGGTC[A/G]CCTTCTGAGTCCCCACACCATAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD19P PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD19P - ankyrin repeat domain 19, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF484 - zinc finger protein 484 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007101.2 | 2666 | Silent Mutation | GGC,GGT | G,G 808 | NP_001007102.1 | |
NM_001261458.1 | 2666 | Silent Mutation | GGC,GGT | G,G 846 | NP_001248387.1 | |
NM_001261459.1 | 2666 | Silent Mutation | GGC,GGT | G,G 808 | NP_001248388.1 | |
NM_001261460.1 | 2666 | Silent Mutation | GGC,GGT | G,G 808 | NP_001248389.1 | |
NM_031486.2 | 2666 | Silent Mutation | GGC,GGT | G,G 844 | NP_113674.1 | |
XM_006717299.3 | 2666 | Silent Mutation | GGC,GGT | G,G 823 | XP_006717362.1 | |
XM_011519073.2 | 2666 | Silent Mutation | GGC,GGT | G,G 876 | XP_011517375.1 | |
XM_011519074.2 | 2666 | Silent Mutation | GGC,GGT | G,G 861 | XP_011517376.1 | |
XM_011519075.2 | 2666 | Silent Mutation | GGC,GGT | G,G 859 | XP_011517377.1 | |
XM_017015178.1 | 2666 | Silent Mutation | GGC,GGT | G,G 823 | XP_016870667.1 | |
XM_017015179.1 | 2666 | Silent Mutation | GGC,GGT | G,G 823 | XP_016870668.1 | |
XM_017015180.1 | 2666 | Silent Mutation | GGC,GGT | G,G 823 | XP_016870669.1 | |
XM_017015181.1 | 2666 | Silent Mutation | GGC,GGT | G,G 808 | XP_016870670.1 |