Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGGAGGTGACTGCCTGGTGTA[C/T]ACTTTGAAGAGGGGTTTTAATCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGD3 - FYVE, RhoGEF and PH domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083536.1 | Intron | NP_001077005.1 | ||||
NM_001286993.1 | Intron | NP_001273922.1 | ||||
NM_033086.2 | Intron | NP_149077.2 | ||||
XM_017015274.1 | Intron | XP_016870763.1 |