Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAGGGAGCCCACACCTCAGAGC[A/G]AAAGAAGAGAACCAGTACAGAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601309 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTCH1 - patched 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000264.3 | 5867 | UTR 3 | NP_000255.2 | |||
NM_001083602.1 | 5867 | UTR 3 | NP_001077071.1 | |||
NM_001083603.1 | 5867 | UTR 3 | NP_001077072.1 | |||
NM_001083604.1 | 5867 | UTR 3 | NP_001077073.1 | |||
NM_001083605.1 | 5867 | UTR 3 | NP_001077074.1 | |||
NM_001083606.1 | 5867 | UTR 3 | NP_001077075.1 | |||
NM_001083607.1 | 5867 | UTR 3 | NP_001077076.1 |