Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATTGTTTTATATTGATATAAC[C/T]TAGAGAATGTTCCAGGTTACCCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602159 MIM: 609871 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CORO2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CORO2A - coronin 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D2 - TBC1 domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267571.1 | 3277 | UTR 3 | NP_001254500.1 | |||
NM_001267572.1 | 3277 | UTR 3 | NP_001254501.1 | |||
NM_018421.3 | 3277 | UTR 3 | NP_060891.3 | |||
XM_011518843.1 | 3277 | UTR 3 | XP_011517145.1 | |||
XM_011518844.2 | 3277 | Intron | XP_011517146.1 | |||
XM_017014899.1 | 3277 | UTR 3 | XP_016870388.1 |