Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACGATCTAGAACTTACACATTA[C/T]GTGCTGCCTTTGCACATGCCCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKS6 - ankyrin repeat and sterile alpha motif domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173551.4 | 6291 | UTR 3 | NP_775822.3 | |||
XM_005251793.4 | 6291 | Intron | XP_005251850.1 | |||
XM_005251794.4 | 6291 | Intron | XP_005251851.1 | |||
XM_006716998.3 | 6291 | Intron | XP_006717061.1 | |||
XM_006716999.2 | 6291 | Intron | XP_006717062.1 | |||
XM_011518357.2 | 6291 | Intron | XP_011516659.1 | |||
XM_017014445.1 | 6291 | Intron | XP_016869934.1 | |||
XM_017014446.1 | 6291 | Intron | XP_016869935.1 | |||
XM_017014447.1 | 6291 | Intron | XP_016869936.1 |