Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAAGCATGTTCCTGTGCAGCTGC[A/G]TGTGGGATCACCTCTCCTCCCCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602938 | ||||||||||||||||||||
Literature Links: |
BAAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAAT - bile acid-CoA:amino acid N-acyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127610.1 | 1383 | Silent Mutation | CAC,CAT | H,H 391 | NP_001121082.1 | |
NM_001701.3 | 1383 | Silent Mutation | CAC,CAT | H,H 391 | NP_001692.1 |