Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCAGGCACTGCTGAACATGCCA[A/G]CCACCAAGTCAACTGCCACCCGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605835 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM2 - transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135820.1 | 4935 | UTR 3 | NP_001129292.1 | |||
NM_013390.2 | 4935 | UTR 3 | NP_037522.1 | |||
XM_005251869.4 | 4935 | UTR 3 | XP_005251926.1 |