Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACGAGGGCCCCGGGCGGACGCCG[C/G]AGAGCGCCGAGGAGGCTTTCCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DNAJC25 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC25 - DnaJ heat shock protein family (Hsp40) member C25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015882.2 | 339 | Missense Mutation | CAG,GAG | Q,E 95 | NP_001015882.2 |
DNAJC25-GNG10 - DNAJC25-GNG10 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004125.3 | 339 | Missense Mutation | CAG,GAG | Q,E 95 | NP_004116.2 |