Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTAGGGCCTTGGAACTGATGAAG[A/G]TACTCTAATTGAGATTTTGGCATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151690 | ||||||||||||||||||||
Literature Links: |
ANXA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANXA1 - annexin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000700.2 | 558 | Missense Mutation | GAT,GGT | D,G 135 | NP_000691.1 | |
XM_011518609.1 | 558 | Missense Mutation | GAT,GGT | D,G 135 | XP_011516911.1 | |
XM_017014657.1 | 558 | Missense Mutation | GAT,GGT | D,G 146 | XP_016870146.1 |