Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGAAGGGGCTCTTCAGGTTGCCT[C/T]GCTGACCCATCTTGTCAGTCCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTGS1 - prostaglandin-endoperoxide synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000962.3 | 1392 | Intron | NP_000953.2 | |||
NM_001271164.1 | 1392 | Intron | NP_001258093.1 | |||
NM_001271165.1 | 1392 | Intron | NP_001258094.1 | |||
NM_001271166.1 | 1392 | Intron | NP_001258095.1 | |||
NM_001271367.1 | 1392 | Intron | NP_001258296.1 | |||
NM_001271368.1 | 1392 | Intron | NP_001258297.1 | |||
NM_080591.2 | 1392 | Intron | NP_542158.1 | |||
XM_005252105.3 | 1392 | Intron | XP_005252162.1 | |||
XM_011518875.2 | 1392 | Intron | XP_011517177.1 | |||
XM_011518876.2 | 1392 | UTR 5 | XP_011517178.1 | |||
XM_017014929.1 | 1392 | Intron | XP_016870418.1 |