Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTAATATGGAGATTTTCACCT[C/T]TCTGGAATCATTCAGTAAAAATTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605976 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBTB26 - zinc finger and BTB domain containing 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304363.1 | 288 | Missense Mutation | AAG,GAG | K,E 69 | NP_001291292.1 | |
NM_001304364.1 | 288 | Missense Mutation | AAG,GAG | K,E 69 | NP_001291293.1 | |
NM_020924.3 | 288 | Missense Mutation | AAG,GAG | K,E 69 | NP_065975.1 |
ZBTB6 - zinc finger and BTB domain containing 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |