Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAGCAAAACCTAGGGCTTAAGA[C/T]GTCAAAATCTTCCAACAGTTCTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612725 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP6C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP6C - protein phosphatase 6 catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123355.1 | 3599 | UTR 3 | NP_001116827.1 | |||
NM_001123369.1 | 3599 | UTR 3 | NP_001116841.1 | |||
NM_002721.4 | 3599 | UTR 3 | NP_002712.1 | |||
XM_011518847.2 | 3599 | UTR 3 | XP_011517149.1 |
SCAI - suppressor of cancer cell invasion | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |