Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTTGCCTGCTCTTGAGTCGCTT[A/G]CTGATGAGGTACATGGCCGCTGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608152 | ||||||||||||||||||||
Literature Links: |
PTGES2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTGES2 - prostaglandin E synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256335.1 | 899 | Silent Mutation | AGC,AGT | S,S 99 | NP_001243264.1 | |
NM_025072.6 | 899 | Silent Mutation | AGC,AGT | S,S 290 | NP_079348.1 | |
NM_198938.2 | 899 | Silent Mutation | AGC,AGT | S,S 99 | NP_945176.1 | |
XM_005252239.2 | 899 | Silent Mutation | AGC,AGT | S,S 99 | XP_005252296.1 | |
XM_011519050.1 | 899 | Silent Mutation | AGC,AGT | S,S 99 | XP_011517352.1 | |
XM_011519051.2 | 899 | Silent Mutation | AGC,AGT | S,S 99 | XP_011517353.1 |
PTGES2-AS1 - PTGES2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A25-AS1 - SLC25A25 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |