Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGACCCCGTGAGAGGCCGTGTAT[A/G]TGTGAGGTCGGCCTGTGTTCACCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf50 - chromosome 9 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NTMT1 - N-terminal Xaa-Pro-Lys N-methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286796.1 | Intron | NP_001273725.1 | ||||
NM_001286797.1 | Intron | NP_001273726.1 | ||||
NM_001286798.1 | Intron | NP_001273727.1 | ||||
NM_001286799.1 | Intron | NP_001273728.1 | ||||
NM_001286800.1 | Intron | NP_001273729.1 | ||||
NM_001286801.1 | Intron | NP_001273730.1 | ||||
NM_001286802.1 | Intron | NP_001273731.1 | ||||
NM_001286803.1 | Intron | NP_001273732.1 | ||||
NM_014064.3 | Intron | NP_054783.2 | ||||
XM_005251939.3 | Intron | XP_005251996.1 | ||||
XM_017014642.1 | Intron | XP_016870131.1 |