Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGTGTGCTGAGCACCCATTTCC[C/T]CACATAAAATTTTTAATAGTCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615251 | ||||||||||||||||||||
Literature Links: |
ZBED5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBED5 - zinc finger BED-type containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143667.1 | 1749 | Missense Mutation | GAG,GGG | E,G 417 | NP_001137139.1 | |
NM_021211.3 | 1749 | Missense Mutation | GAG,GGG | E,G 417 | NP_067034.2 |
ZBED5-AS1 - ZBED5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |