Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCGCCAATTCTGTACCTCCCGGG[G/T]CTGAGGAAGAGCCGAGGTGACTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614460 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP47 - ubiquitin specific peptidase 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282659.1 | 397 | UTR 5 | NP_001269588.1 | |||
NM_017944.3 | 397 | UTR 5 | NP_060414.3 | |||
XM_005252997.1 | 397 | UTR 5 | XP_005253054.1 | |||
XM_011520196.1 | 397 | Intron | XP_011518498.1 | |||
XM_017017953.1 | 397 | UTR 5 | XP_016873442.1 | |||
XM_017017954.1 | 397 | Intron | XP_016873443.1 |