Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGCCCTGACGGAAGCATCCTTC[A/C]CTGTGATCCACCTCAGCAATGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607228 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRGPRX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRGPRX2 - MAS related GPR family member X2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303615.1 | 1027 | Missense Mutation | AGG,AGT | R,S 313 | NP_001290544.1 | |
NM_054030.3 | 1027 | Missense Mutation | AGG,AGT | R,S 313 | NP_473371.1 |