Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAAAATTCTGTAGCAATTGTTC[A/C]GCACCTTCCTGTCCTGGAGACTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1549L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1549L - KIAA1549 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012194.2 | Intron | NP_036326.2 | ||||
XM_005252847.3 | Intron | XP_005252904.2 | ||||
XM_005252848.3 | Intron | XP_005252905.1 | ||||
XM_011519969.2 | Intron | XP_011518271.1 | ||||
XM_011519970.2 | Intron | XP_011518272.1 | ||||
XM_017017484.1 | Intron | XP_016872973.1 | ||||
XM_017017485.1 | Intron | XP_016872974.1 | ||||
XM_017017486.1 | Intron | XP_016872975.1 |