Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCTGGGGCTCCTGGGTGCCTCC[C/T]TCTATCTCTGTGTGATGTCTCTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107271 MIM: 609089 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD59 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD59 - CD59 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO3 - F-box protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012175.3 | 1454 | Intron | NP_036307.2 | |||
NM_033406.2 | 1454 | Intron | NP_208385.1 | |||
XM_011519980.1 | 1454 | Missense Mutation | AGG,GGG | R,G 456 | XP_011518282.1 | |
XM_011519981.2 | 1454 | Intron | XP_011518283.1 | |||
XM_011519982.1 | 1454 | Missense Mutation | AGG,GGG | R,G 294 | XP_011518284.1 | |
XM_017017533.1 | 1454 | Intron | XP_016873022.1 |