Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATGGTGGAGCTGCAGGTAACC[A/G]CCACATTGGAGGTCACAGAGTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMO2 - LIM domain only 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142315.1 | Intron | NP_001135787.1 | ||||
NM_001142316.1 | Intron | NP_001135788.1 | ||||
NM_005574.3 | Intron | NP_005565.2 | ||||
XM_005252921.2 | Intron | XP_005252978.1 | ||||
XM_017017727.1 | Intron | XP_016873216.1 | ||||
XM_017017728.1 | Intron | XP_016873217.1 | ||||
XM_017017729.1 | Intron | XP_016873218.1 | ||||
XM_017017730.1 | Intron | XP_016873219.1 | ||||
XM_017017731.1 | Intron | XP_016873220.1 | ||||
XM_017017732.1 | Intron | XP_016873221.1 | ||||
XM_017017733.1 | Intron | XP_016873222.1 |