Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTAAGTTAGAGCATCTCTTCCTG[A/C]AATGTGATACCCATGATAAAAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600300 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC1A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC1A2 - solute carrier family 1 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195728.2 | 7815 | UTR 3 | NP_001182657.1 | |||
NM_001252652.1 | 7815 | UTR 3 | NP_001239581.1 | |||
NM_004171.3 | 7815 | UTR 3 | NP_004162.2 | |||
XM_005253067.1 | 7815 | UTR 3 | XP_005253124.1 | |||
XM_011520285.2 | 7815 | UTR 3 | XP_011518587.1 | |||
XM_017018136.1 | 7815 | UTR 3 | XP_016873625.1 | |||
XM_017018137.1 | 7815 | UTR 3 | XP_016873626.1 | |||
XM_017018138.1 | 7815 | UTR 3 | XP_016873627.1 | |||
XM_017018139.1 | 7815 | UTR 3 | XP_016873628.1 |