Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCACACAGCAGCGAGTCCACCA[C/T]ACTGACCACCCCAGAGCGCAGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAMR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAMR1 - peptidase domain containing associated with muscle regeneration 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001991.2 | 2367 | Missense Mutation | ATG,GTG | M,V 624 | NP_001001991.1 | |
NM_001282675.1 | 2367 | Missense Mutation | ATG,GTG | M,V 584 | NP_001269604.1 | |
NM_001282676.1 | 2367 | Missense Mutation | ATG,GTG | M,V 513 | NP_001269605.1 | |
NM_015430.3 | 2367 | Missense Mutation | ATG,GTG | M,V 641 | NP_056245.2 |