Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAGCGACTCCGAGTCGGCTTCC[A/G]CCTCGTCCAGCGGCTCCGAGCGCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601441 | ||||||||||||||||||||
Literature Links: |
DGKZ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DGKZ - diacylglycerol kinase zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105540.1 | 312 | Intron | NP_001099010.1 | |||
NM_001199266.1 | 312 | Missense Mutation | ACC,GCC | T,A 20 | NP_001186195.1 | |
NM_001199267.1 | 312 | Missense Mutation | ACC,GCC | T,A 20 | NP_001186196.1 | |
NM_001199268.1 | 312 | Missense Mutation | ACC,GCC | T,A 20 | NP_001186197.1 | |
NM_003646.3 | 312 | Missense Mutation | ACC,GCC | T,A 20 | NP_003637.2 | |
NM_201532.2 | 312 | Intron | NP_963290.1 | |||
NM_201533.3 | 312 | Intron | NP_963291.2 | |||
XM_005253181.3 | 312 | Intron | XP_005253238.1 | |||
XM_005253182.3 | 312 | Intron | XP_005253239.1 | |||
XM_006718354.1 | 312 | Intron | XP_006718417.1 | |||
XM_006718355.2 | 312 | Intron | XP_006718418.1 | |||
XM_011520422.1 | 312 | Intron | XP_011518724.1 | |||
XM_011520423.1 | 312 | Intron | XP_011518725.1 | |||
XM_017018455.1 | 312 | Intron | XP_016873944.1 | |||
XM_017018456.1 | 312 | Intron | XP_016873945.1 | |||
XM_017018457.1 | 312 | Intron | XP_016873946.1 |