Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGCCAATGAATTAACACAACTCT[G/T]GATTCCATCTCAAATCCCTCTCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142290 MIM: 605493 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPX - hemopexin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM3 - tripartite motif containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001248006.1 | Intron | NP_001234935.1 | ||||
NM_001248007.1 | Intron | NP_001234936.1 | ||||
NM_006458.3 | Intron | NP_006449.2 | ||||
NM_033278.3 | Intron | NP_150594.2 | ||||
XM_011519844.1 | Intron | XP_011518146.1 | ||||
XM_011519845.2 | Intron | XP_011518147.1 | ||||
XM_017017098.1 | Intron | XP_016872587.1 | ||||
XM_017017099.1 | Intron | XP_016872588.1 |