Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTGGGTTGAGAAAGCCAAGAGG[A/G]GGGCGGCCACTAAGGATCCTGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603057 MIM: 602366 MIM: 600475 MIM: 607998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCHS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCHS1 - dachsous cadherin-related 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ILK - integrin linked kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF10 - TATA-box binding protein associated factor 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPP1 - tripeptidyl peptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000391.3 | 1555 | Silent Mutation | CCC,CCT | P,P 498 | NP_000382.3 |