Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAATGAGGCCAGATCAGGAGGGT[C/T]GTGGATGTCAACAGTTGTGGCGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605016 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR2D3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR2D3 - olfactory receptor family 2 subfamily D member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF215 - zinc finger protein 215 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013250.2 | Intron | NP_037382.2 | ||||
XM_006718311.2 | Intron | XP_006718374.1 |