Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGGCTGAGGACAGTGTGTGAC[A/G]GTCGCCGCCAAAAGCAGTTCTACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608213 MIM: 609302 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEND1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEND1 - cell cycle exit and neuronal differentiation 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016564.3 | 1420 | UTR 3 | NP_057648.2 |
LOC171391 - uncharacterized LOC171391 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDDC1 - Parkinson disease 7 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A22 - solute carrier family 25 member 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |