Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGTAGAAGCTTCCTTTTGTGTT[C/T]CTGCTATATTATCTTTGCCCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609662 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP10 - NLR family pyrin domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176821.3 | 2125 | Missense Mutation | GAA,GGA | E,G 633 | NP_789791.1 | |
XM_011520043.2 | 2125 | Missense Mutation | GAA,GGA | E,G 633 | XP_011518345.1 |