Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTGTCAGCCACTCTGTATAACT[C/T]TCCTCGAATATTTACCATGTTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609191 MIM: 140750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKIP1 - A-kinase interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206646.1 | 2043 | Intron | NP_001193575.1 | |||
NM_001206647.1 | 2043 | Intron | NP_001193576.1 | |||
NM_001206648.1 | 2043 | Intron | NP_001193577.1 | |||
NM_020642.3 | 2043 | Intron | NP_065693.2 | |||
XM_017018011.1 | 2043 | Intron | XP_016873500.1 | |||
XM_017018012.1 | 2043 | Intron | XP_016873501.1 |
C11orf16 - chromosome 11 open reading frame 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020643.2 | 2043 | UTR 3 | NP_065694.2 | |||
XM_017018013.1 | 2043 | UTR 3 | XP_016873502.1 |
ST5 - suppression of tumorigenicity 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |