Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATGAGGAACGTGGCTCAGTGAA[A/G]ATGGTGACTTGATATTTAACCGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GYS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GYS2 - glycogen synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021957.3 | 2309 | Missense Mutation | TCT,TTT | S,F 685 | NP_068776.2 | |
XM_006719063.3 | 2309 | Missense Mutation | TCT,TTT | S,F 608 | XP_006719126.1 | |
XM_017019245.1 | 2309 | Intron | XP_016874734.1 |
SPX - spexin hormone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |