Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACATACGTCGTAAACTGCAGAC[A/G]TCCCAGTGTCATAGGCAAGGAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601103 | ||||||||||||||||||||
Literature Links: |
MFAP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MFAP5 - microfibrillar associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297709.1 | Intron | NP_001284638.1 | ||||
NM_001297710.1 | Intron | NP_001284639.1 | ||||
NM_001297711.1 | Intron | NP_001284640.1 | ||||
NM_001297712.1 | Intron | NP_001284641.1 | ||||
NM_003480.3 | Intron | NP_003471.1 |