Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGAATTTCTCGAACTAATGTATA[G/C]AAGGCATCATCAACACCCTGAAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 190070 | ||||||||||||||||||||
Literature Links: |
KRAS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRAS - KRAS proto-oncogene, GTPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004985.4 | 790 | Missense Mutation | TTC,TTG | F,L 156 | NP_004976.2 | |
NM_033360.3 | 790 | UTR 3 | NP_203524.1 | |||
XM_006719069.3 | 790 | UTR 3 | XP_006719132.1 | |||
XM_011520653.2 | 790 | Intron | XP_011518955.1 | |||
XM_017019292.1 | 790 | Intron | XP_016874781.1 | |||
XM_017019293.1 | 790 | Missense Mutation | TTC,TTG | F,L 195 | XP_016874782.1 |
LYRM5 - LYR motif containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |