Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTGCTGAGGCAGAGGGTAGAC[A/G]TGTACTGGCAGTATGGTGGCTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610375 | ||||||||||||||||||||
Literature Links: |
CAPRIN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPRIN2 - caprin family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002259.2 | 2891 | Silent Mutation | CAC,CAT | H,H 988 | NP_001002259.1 | |
NM_001206856.2 | 2891 | UTR 3 | NP_001193785.1 | |||
NM_001319842.1 | 2891 | Silent Mutation | CAC,CAT | H,H 654 | NP_001306771.1 | |
NM_001319843.1 | 2891 | Silent Mutation | CAC,CAT | H,H 987 | NP_001306772.1 | |
NM_001319844.1 | 2891 | Silent Mutation | CAC,CAT | H,H 872 | NP_001306773.1 | |
NM_001319845.1 | 2891 | Silent Mutation | CAC,CAT | H,H 907 | NP_001306774.1 | |
NM_001319846.1 | 2891 | Silent Mutation | CAC,CAT | H,H 907 | NP_001306775.1 | |
NM_023925.4 | 2891 | Silent Mutation | CAC,CAT | H,H 938 | NP_076414.2 | |
NM_032156.4 | 2891 | UTR 3 | NP_115532.3 | |||
XM_006719138.3 | 2891 | Silent Mutation | CAC,CAT | H,H 967 | XP_006719201.1 | |
XM_006719139.3 | 2891 | Silent Mutation | CAC,CAT | H,H 953 | XP_006719202.1 | |
XM_006719140.3 | 2891 | Silent Mutation | CAC,CAT | H,H 952 | XP_006719203.1 | |
XM_006719142.3 | 2891 | Silent Mutation | CAC,CAT | H,H 918 | XP_006719205.1 | |
XM_006719144.3 | 2891 | Silent Mutation | CAC,CAT | H,H 904 | XP_006719207.1 | |
XM_006719145.3 | 2891 | UTR 3 | XP_006719208.1 | |||
XM_006719146.3 | 2891 | UTR 3 | XP_006719209.1 | |||
XM_006719147.3 | 2891 | UTR 3 | XP_006719210.1 | |||
XM_017019851.1 | 2891 | Silent Mutation | CAC,CAT | H,H 932 | XP_016875340.1 | |
XM_017019852.1 | 2891 | Silent Mutation | CAC,CAT | H,H 906 | XP_016875341.1 | |
XM_017019853.1 | 2891 | Silent Mutation | CAC,CAT | H,H 906 | XP_016875342.1 | |
XM_017019854.1 | 2891 | Silent Mutation | CAC,CAT | H,H 903 | XP_016875343.1 | |
XM_017019855.1 | 2891 | Silent Mutation | CAC,CAT | H,H 886 | XP_016875344.1 | |
XM_017019856.1 | 2891 | Silent Mutation | CAC,CAT | H,H 872 | XP_016875345.1 | |
XM_017019857.1 | 2891 | Silent Mutation | CAC,CAT | H,H 871 | XP_016875346.1 | |
XM_017019858.1 | 2891 | Silent Mutation | CAC,CAT | H,H 871 | XP_016875347.1 | |
XM_017019859.1 | 2891 | Silent Mutation | CAC,CAT | H,H 823 | XP_016875348.1 | |
XM_017019860.1 | 2891 | UTR 3 | XP_016875349.1 | |||
XM_017019861.1 | 2891 | Silent Mutation | CAC,CAT | H,H 822 | XP_016875350.1 | |
XM_017019862.1 | 2891 | UTR 3 | XP_016875351.1 | |||
XM_017019863.1 | 2891 | UTR 3 | XP_016875352.1 | |||
XM_017019864.1 | 2891 | Intron | XP_016875353.1 | |||
XM_017019865.1 | 2891 | UTR 3 | XP_016875354.1 | |||
XM_017019866.1 | 2891 | UTR 3 | XP_016875355.1 | |||
XM_017019867.1 | 2891 | UTR 3 | XP_016875356.1 | |||
XM_017019868.1 | 2891 | UTR 3 | XP_016875357.1 | |||
XM_017019869.1 | 2891 | UTR 3 | XP_016875358.1 | |||
XM_017019870.1 | 2891 | UTR 3 | XP_016875359.1 | |||
XM_017019871.1 | 2891 | UTR 3 | XP_016875360.1 | |||
XM_017019872.1 | 2891 | UTR 3 | XP_016875361.1 | |||
XM_017019873.1 | 2891 | UTR 3 | XP_016875362.1 | |||
XM_017019874.1 | 2891 | Silent Mutation | CAC,CAT | H,H 654 | XP_016875363.1 | |
XM_017019875.1 | 2891 | UTR 3 | XP_016875364.1 | |||
XM_017019876.1 | 2891 | Silent Mutation | CAC,CAT | H,H 634 | XP_016875365.1 | |
XM_017019877.1 | 2891 | Silent Mutation | CAC,CAT | H,H 620 | XP_016875366.1 | |
XM_017019878.1 | 2891 | Silent Mutation | CAC,CAT | H,H 619 | XP_016875367.1 | |
XM_017019879.1 | 2891 | Silent Mutation | CAC,CAT | H,H 599 | XP_016875368.1 | |
XM_017019880.1 | 2891 | Silent Mutation | CAC,CAT | H,H 550 | XP_016875369.1 | |
XM_017019881.1 | 2891 | UTR 3 | XP_016875370.1 | |||
XM_017019882.1 | 2891 | UTR 3 | XP_016875371.1 | |||
XM_017019883.1 | 2891 | UTR 3 | XP_016875372.1 |