Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGACTGTCTTCTGCCCTCACAA[C/T]TTGCCTTTTCCATGCTTTTCCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607467 | ||||||||||||||||||||
Literature Links: |
CLECL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLECL1 - C-type lectin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253750.1 | 1066 | UTR 3 | NP_001240679.1 | |||
NM_001267701.1 | 1066 | UTR 3 | NP_001254630.1 | |||
NM_172004.3 | 1066 | Intron | NP_742001.1 | |||
XM_011520574.2 | 1066 | Intron | XP_011518876.1 | |||
XM_017018885.1 | 1066 | Intron | XP_016874374.1 |