Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGACCGAAGCGTCCCAGCCGATTG[C/T]GGAAGAGGAGGAAACTAAAACATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615428 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX47 - DEAD-box helicase 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016355.3 | 75 | Missense Mutation | GCG,GTG | A,V 18 | NP_057439.2 | |
NM_201224.1 | 75 | Missense Mutation | GCG,GTG | A,V 18 | NP_957518.1 |