Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGCCGGAATCTACAGGAAACGA[A/G]AAGGTAAGCTTTTGAAATGGCCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KMT5A - lysine methyltransferase 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324504.1 | 585 | Intron | NP_001311433.1 | |||
NM_001324505.1 | 585 | Intron | NP_001311434.1 | |||
NM_001324506.1 | 585 | Intron | NP_001311435.1 | |||
NM_020382.4 | 585 | Intron | NP_065115.3 | |||
XM_006719394.3 | 585 | Intron | XP_006719457.1 | |||
XM_011538333.2 | 585 | Missense Mutation | AAA,GAA | K,E 39 | XP_011536635.1 |