Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCCTAGTGTTCCACACCAACAG[A/G]GGACCTATTAAGTTCAATGTATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601179 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAN - RAN, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300796.1 | 257 | UTR 5 | NP_001287725.1 | |||
NM_001300797.1 | 257 | UTR 5 | NP_001287726.1 | |||
NM_006325.4 | 257 | Silent Mutation | AGA,AGG | R,R 56 | NP_006316.1 | |
XM_017019772.1 | 257 | Silent Mutation | AGA,AGG | R,R 56 | XP_016875261.1 | |
XM_017019773.1 | 257 | UTR 5 | XP_016875262.1 |