Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATGGAAGGCGTGGCAGGAGTC[C/T]GGTCGCCGGCGGAGCTTCTGGGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607946 MIM: 608171 | ||||||||||||||||||||
Literature Links: |
ADIPOR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADIPOR2 - adiponectin receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CACNA2D4 - calcium voltage-gated channel auxiliary subunit alpha2delta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172364.4 | 7352 | Silent Mutation | CCA,CCG | P,P 1094 | NP_758952.4 | |
XM_011521041.2 | 7352 | Silent Mutation | CCA,CCG | P,P 1073 | XP_011519343.1 |