Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCCTGGAGTCAGGGTTGTAGGT[A/G]GGGGGTGAGAAGGAAAGGCTGTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRACR2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRACR2A - calcium release activated channel regulator 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144958.1 | 2325 | Intron | NP_001138430.1 | |||
NM_032680.3 | 2325 | Intron | NP_116069.1 | |||
XM_006719021.3 | 2325 | UTR 3 | XP_006719084.1 | |||
XM_011521034.2 | 2325 | UTR 3 | XP_011519336.1 | |||
XM_011521036.2 | 2325 | UTR 3 | XP_011519338.1 | |||
XM_011521037.2 | 2325 | UTR 3 | XP_011519339.1 | |||
XM_011521038.2 | 2325 | UTR 3 | XP_011519340.1 |