Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATCGGTGGGCGTTGGGAGCATC[A/G]GCGGAGATGCACGCAATGATAAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120580 | ||||||||||||||||||||
Literature Links: |
C1S PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1S - complement component 1, s subcomponent | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001734.3 | 42 | Intron | NP_001725.1 | |||
NM_201442.2 | 42 | Intron | NP_958850.1 | |||
XM_005253760.1 | 42 | UTR 5 | XP_005253817.1 |