Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAACAAAAAACAGCCTTTTTTC[A/G]TGGTCTTAGATGATTATTTCCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600392 MIM: 605232 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD52 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAD52 - RAD52 homolog, DNA repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297419.1 | 2279 | UTR 3 | NP_001284348.1 | |||
NM_001297420.1 | 2279 | Intron | NP_001284349.1 | |||
NM_001297421.1 | 2279 | UTR 3 | NP_001284350.1 | |||
NM_001297422.1 | 2279 | Intron | NP_001284351.1 | |||
NM_134424.3 | 2279 | UTR 3 | NP_602296.2 | |||
XM_005253720.4 | 2279 | UTR 3 | XP_005253777.1 | |||
XM_005253721.2 | 2279 | UTR 3 | XP_005253778.1 | |||
XM_011520990.2 | 2279 | UTR 3 | XP_011519292.1 | |||
XM_011520991.2 | 2279 | UTR 3 | XP_011519293.1 | |||
XM_011520993.2 | 2279 | Intron | XP_011519295.1 | |||
XM_011520995.2 | 2279 | UTR 3 | XP_011519297.1 | |||
XM_017019769.1 | 2279 | UTR 3 | XP_016875258.1 | |||
XM_017019770.1 | 2279 | UTR 3 | XP_016875259.1 | |||
XM_017019771.1 | 2279 | Intron | XP_016875260.1 |
WNK1 - WNK lysine deficient protein kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |